Learn More
Abcam AY 9944 dihydrochloride, Hedgehog signaling inhibitor, 50MG
Supplier:  Abcam AB14588050MG
Encompass_Preferred
MW 464.3 g/mol, Purity >99%. Hedgehog signaling inhibitor. Potent Δ7-dehydrocholesterol reductase inhibitor (IC₅₀ = 13 nM). Inhibits cholesterol esterification and biosynthesis, and shows teratogenic effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5043