Learn More
Selleck Chemical LLC AZ-33 S0108-25MG
SDP
Supplier:  Selleck Chemical LLC S010825MG
Encompass_Preferred
AZ-33 (LDHA Inhibitor 33) is a potent inhibitor of lactate dehydrogenase A (LDHA) with IC50 of 0 5 M LDHA is the key enzyme involved in anaerobic glycolysis which is frequently deregulated in human malignancies
Catalog No. 50-000-40190