Learn More
Medchemexpress LLC AZ-5104 | 1421373-98-9 | 99.91% | 200 MG

Supplier: Medchemexpress LLC HYB0793200MG
AZ-5104 is an active, demethylated metabolite of AZD 9291 and functions as an EGFR inhibitor. It demonstrates inhibitory effects on EGFR phosphorylation across various cell lines, targeting both mutant and wild-type EGFR.
- Inhibits EGFR with specific IC50 values across multiple targets
- Effective in tumor shrinkage in C/L858R and C/L+T mice
- Shows minimal off-target activity against non-HER family kinases
- Potentially targets both HER2 and HER4 kinase activity
- Store powder at -20°C for 3 years or 4°C for 2 years
- Store in solvent at -80°C for 2 years or -20°C for 1 year
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.