Learn More
Medchemexpress LLC HY-11004 50mg , AZ 628 CAS:878739-06-1 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY1100450MG
Encompass_Preferred
Medchemexpress, HY-11004 50mg AZ 628 CAS:878739-06-1 Formula:C27H25N5O2 IC50: 105 nM (B-Raf), 34 nM (B-RafV600E), 29 nM (c-Raf-1) Purity:98% AZ628 is a pan-Raf kinase inhibitor with IC 50 s of 105, 34 and 29 nM for B-Raf , B-RafV600E , and c-Raf-1 , respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3669
 Restricted Item