Learn More
Medchemexpress LLC AZ 628 | 878739-06-1 | 99.4% | 451.52 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1100410MG
Encompass_Preferred
AZ 628 is a pan-Raf kinase inhibitor with IC50s of 105, 34, and 29 nM for B-Raf, B-RafV600E, and c-Raf-1, respectively. It reduces the activities of preactivated B-Raf, B-RafV600E, and c-Raf-1 in in vitro kinase assays.
  • Inhibits activation of several tyrosine protein kinases including VEGFR2, DDR2, Lyn, Flt1, and FMS.
  • Inhibits anchorage-dependent and -independent growth.
  • Causes cell cycle arrest.
  • Induces apoptosis in colon and melanoma cell lines harboring the B-RafV600E mutation.
  • Suppresses growth in cells expressing K-RASG13D.
  • Selectively affects viability in K-RAS mutant cells by suppressing MEK and ERK phosphorylation.
Catalog No. 50-003-45468