Learn More
Medchemexpress LLC AZ'9567 | 3056570-19-2 | 99.2% | 447.44 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1623535MG
Encompass_Preferred
AZ9567 is identified as a potent inhibitor of MAT2A. It has demonstrated antiproliferative activity against MTAP KO HCT116 cells, with a pIC50 of 8.9.
  • Potent MAT2A inhibitor.
  • Exhibits antiproliferative activity against MTAP KO HCT116 cells.
  • Available in solid form (white to off-white appearance).
  • High purity of 99.21%.
Catalog No. 50-003-92762