Learn More
Selleck Chemical LLC AZ 960 S2214-1g
SDP
Supplier:  Selleck Chemical LLC S22141G
Encompass_Preferred
AZ 960 is a novel ATP competitive JAK2 inhibitor with IC50 and Ki of 3 nM and 0 45 nM 3-fold selectivity of AZ960 for JAK2 over JAK3 AZ 960 induces apoptosis and growth arrest
Catalog No. 50-311-2777