Learn More
Medchemexpress LLC Az-dyrk1b-33 | 1679330-37-0 | 100.0% | 300.36 g/mol | C19H16N4 | 1 ML

Supplier: Medchemexpress LLC HY11739110MM 1ML
AZ-Dyrk1B-33 is a potent, selective inhibitor of Dyrk1B kinase supplied as a 10 mM solution in DMSO (1 mL) for research use. It exhibits an enzymatic IC50 of 7 nM and cellular activity inhibiting pS421 with an IC50 ≈ 0.192 μM; purity is reported at 99.95%.
- Potent Dyrk1B inhibition (enzymatic IC50 7 nM).
- Shows cellular activity (pS421 IC50 ≈ 0.192 μM in COS-1 cells).
- Supplied as a 10 mM solution in DMSO, 1 mL vial.
- High purity (99.95%).
- Molecular weight 300.36 g/mol; formula C19H16N4.
- Intended for in vitro biochemical and cell-based assays; for research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.