Learn More
Medchemexpress LLC Ikarugamycin | 36531-78-9 | 99.9% | 1 MG

Supplier: Medchemexpress LLC HY1197641MG
Ikarugamycin is a macrocyclic antibiotic used as a research reagent to inhibit clathrin-mediated endocytosis (CME) and probe cellular uptake pathways. It is supplied at high purity for use in cellular and biochemical assays.
- Inhibits clathrin-mediated endocytosis for mechanistic studies.
- High purity, 99.9%, suitable for research applications.
- Chemical formula C29H38N2O4 and molecular weight 478.62 g·mol⁻¹.
- Provided in small milligram-scale quantities for laboratory experiments.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.