Learn More
Medchemexpress LLC AZ194 | 2241651-99-8 | 99.8% | C34H31F2N3O3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY14516910MM/1ML
Encompass_Preferred
AZ194 is a first-in-class, orally active inhibitor of CRMP2-Ubc9 interaction and an inhibitor of NaV1.7 with an IC50 of 1.2 μM. It works by blocking SUMOylation of CRMP2, which selectively reduces the amount of surface-expressed NaV1.7, exhibiting antinociceptive effects.
  • First-in-class orally active inhibitor
  • Inhibits CRMP2-Ubc9 interaction
  • Inhibits NaV1.7 (IC50 = 1.2 μM)
  • Blocks SUMOylation of CRMP2
  • Selectively reduces surface-expressed NaV1.7
  • Exhibits antinociceptive effects
  • Provides pain relief in rat models of neuropathic pain
  • Restores mechanical sensitivity
  • Does not affect motor performance
  • Synergizes with commonly used painkillers
Catalog No. 50-003-56380