Learn More
TARGETMOL CHEMICALS INC AZ3146 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2689
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AZ3146 is a selective Mps1 inhibitor with IC50 of ~35 nM. purity: 98%
Catalog No. 50-258-0665