Learn More
Apexbio Technology LLC AZ505 1035227-43-0 10mg
SDP
Supplier:  Apexbio Technology LLC B125510
Encompass_Preferred
AZ505 (CAS 1035227-43-0) is a small molecule identified through high-throughput chemical screening as a potent and selective inhibitor of SET and MYND domain-containing protein 2 (SMYD2) a lysine methyltransferase AZ505 inhibits SMYD2 with an IC50 of 0 12 M and a Ki of 0 3 M It acts as a substrate-competitive inhibitor binding to the peptide substrate recognition pocket of SMYD2 rather than the cofactor site SMYD2 mediates methylation of both histone proteins (H2B H3 and H4) and non-histone substrates such as the tumor suppressors p53 and Rb AZ505 is valuable for investigating SMYD2 function and epigenetic regulatory mechanisms in cancer and other diseases
Catalog No. 50-000-92167