Learn More
Medchemexpress LLC AZ5576 | 2751721-40-9 | 99.9% | C21H24FN3O3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY143584100MG
Encompass_Preferred
AZ5576 is a potent and highly selective CDK9 inhibitor with an IC50 of less than 5 nM. It works by inhibiting the phosphorylation of RNA polymerase II at Ser2, which in turn inhibits transcriptional elongation. This product can be used for research related to hematological malignancies.
  • Potent and highly selective CDK9 inhibitor
  • Inhibits phosphorylation of RNA polymerase II at Ser2
  • Inhibits transcriptional elongation
  • Reduces Mcl-1 and MYC mRNA and protein expressions
  • Inhibits DNA synthesis in DLBCL cells
  • Induces apoptosis in DLBCL cells
  • Soluble in DMSO at 125 mg/mL (requires sonication)
  • Appears as a white to off-white solid
Catalog No. 50-003-54924