Learn More
Medchemexpress LLC HY-119653 1mg Medchemexpress, AZ9482 CAS:1825345-33-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1196531MG
Encompass_Preferred
Medchemexpress, HY-119653 1mg AZ9482 CAS:1825345-33-2 AZ9482 is a triple PARP1/2/6 inhibitor, with IC50 values of 1 nM, 1 nM and 640 nM for PARP1, PARP2 and PARP6, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0930