Learn More
Medchemexpress LLC Azadirachtin (standard) | 11141-17-6 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY12674110MG
Encompass_Preferred
Azadirachtin (Standard) is the analytical standard of Azadirachtin, intended for research and analytical applications. As one of the most promising botanical insecticides, it is widely used for pest control and induces apoptosis in insect cell lines, including Sf9, SL-1, and BTI-Tn-5B1-4. This compound serves as an analytical standard for reference in assays and is commonly utilized in qualitative, quantitative, and methodological research experiments involving HPLC, GC, and MS.
  • Analytical standard
  • Intended for research and analytical applications
  • Used for pest control
  • Induces apoptosis in insect cell lines
  • Utilized in HPLC, GC, and MS research experiments
Catalog No. 50-003-41523