Learn More
Medchemexpress LLC Azamethiphos 10mM 1mL | 35575-96-3 | 324.68 g/mol | C9H10ClN2O5PS | 1 ML

Supplier: Medchemexpress LLC HY11489910MM 1ML
Azamethiphos is an organophosphate insecticide and acetylcholinesterase (AChE) inhibitor supplied as an analytical standard for laboratory research. The product is provided as a ready-to-use 10 mM solution in DMSO (1 mL) and is also available in solid quantities for analytical and toxicology studies. For research use only; not for human use.
- Organophosphate acetylcholinesterase inhibitor.
- Ready-to-use 10 mM solution in DMSO, 1 mL.
- Also available as solid in 100 mg-1 g sizes.
- Purity 96.46% as listed.
- Molecular weight 324.68 g/mol; formula C9H10ClN2O5PS.
- Intended for analytical, assay, and toxicology applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.