Learn More
Cayman Chemical AzasetronhydrochlorIde 100mg
SDP
Supplier:  Cayman Chemical 28383100 MG
Encompass_Preferred
An orally bioavailable 5-HT3 antagonist (Ki = 2.9 nM); selective for 5-HT3 over 5-HT1A and 5-HT2 (IC50 = >10 μM for both), as well as D1, D2, α1-, and α2-adrenergic receptors (IC50s = >10 μM); binds to H1 receptors (IC50 = 4.4 μM); inhibits 5-HT-induced contractions in isolated guinea pig ileum (pA2 = 7.04) and isolated rabbit heart (pA2 = 10.06); inhibits cisplatin-induced emesis in dogs and doxorubicin/cyclophosphamide-induced emesis in ferrets at 1 mg/kg
Catalog No. 50-293-9474