Learn More
Medchemexpress LLC HY-B0256 500mg , Azathioprine CAS:446-86-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0256500MG
Encompass_Preferred
HY-B0256 500mg Azathioprine CAS: 446-86-6 Azathioprine (BW 57-322) is an orally active immunosuppressive agent. Azathioprine can be converted in vivo to the active metabolite 6-mercaptopurine (6-MP). Azathioprine has myelosuppressive effects and induces apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0289