Learn More
Medchemexpress LLC Azd-0284 | 2101291-07-8 | 99.9% | 10 MG

Supplier: Medchemexpress LLC HY12038410MG
AZD-0284 is a selective inverse agonist of the retinoic acid receptor-related orphan receptor γ (RORγ), used as a research tool to study Th17-mediated inflammation and autoimmune disease pathways.
- Selective inverse agonist of RORγ suitable for in vitro and preclinical studies.
- High reported purity (99.9%) for consistent experimental results.
- Available in small-mass formats and DMSO solution for formulation flexibility.
- Documented molecular formula C21H18F6N2O5S and molecular weight 524.43 g/mol.
- Analytical data (e.g., LCMS, COA) reported to support compound identity.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.