Learn More
Medchemexpress LLC AZD-1678 | 942137-41-9 | 98.1% | 352.17 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10951110MM/1ML
Encompass_Preferred
AZD-1678 is a potent CCR4 receptor antagonist, with a pIC50 of 8.6. It is intended for research use only.
  • Potent CCR4 receptor antagonist
  • pIC50 of 8.6
Catalog No. 50-003-45062