Learn More
TARGETMOL CHEMICALS INC AZD 3147 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T362005MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AZD 3147 is an inhibitor of mTORC with IC50s of 40.7 and 5.75 nM for mTORC1 and mTORC2. AZD 3147 shows IC50s of 912 5495 9333 and 6310 nM for PI3K(alpha) PI3K(beta) PI3K(delta) and PI3K(gamma) respectively. purity: 98%
Catalog No. 50-258-0196