Learn More
Selleck Chemical LLC AZD-5462 E1995-100MG
SDP
Supplier:  Selleck Chemical LLC E1995100MG
Encompass_Preferred
AZD-5462(Example 1) is a modulator of RXFP1 the cognate receptor for human relaxin that belongs to the GPCR family 1c number AZD-5462 exhibits anti-fibrotic and anti-inflammatory properties
Catalog No. 50-000-39266
Add to Cart
Request a Quote