Learn More
Medchemexpress LLC AZD-5991 (S-enantiomer) | 2143061-82-7 | 99.8% | 672.26 g/mol | C35H34ClN5O3S2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY101533B10MG
Encompass_Preferred
AZD-5991 (S-enantiomer) is the less active stereoisomer of a small-molecule Mcl-1 inhibitor provided for research use. It is characterized in biochemical binding and inhibition assays and includes reported activity values from FRET and SPR assays. Molecular formula C35H34ClN5O3S2 and molecular weight 672.26 g/mol are documented for identity and calculation purposes.
  • Stereochemically defined S-enantiomer for targeted stereospecific studies.
  • Characterized biochemical activity: IC50 6.3 μM (FRET), Kd 0.98 μM (SPR).
  • High purity suitable for research: 99.83% reported.
  • Molecular formula and molecular weight provided for planning and analysis.
  • Supplied in a small research quantity (10 MG) for preclinical experiments.
Catalog No. 50-002-45813