Learn More
Medchemexpress LLC HY-10422 200mg , AZD-8055 CAS:1009298-09-2 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY10422200MG
Encompass_Preferred
Medchemexpress, HY-10422 200mg AZD-8055 CAS:1009298-09-2 Formula:C25H31N5O4 IC50: 0.8 nM (mTOR) Purity:98% AZD-8055 is a novel ATP-competitive inhibitor of mTOR kinase activity, with an IC 50 of 0.8 nM. AZD-8055 inhibits both mTORC1 and mTORC2 . Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2560