Learn More
Medchemexpress LLC AZD-8835 | 1620576-64-8 | 98.6% | 100 MG
Supplier: Medchemexpress LLC HY12869100MG
AZD-8835 is a potent and selective inhibitor of PI3Kα and PI3Kδ, with IC50s of 6.2 nM and 5.7 nM, respectively. It also potently inhibits commonly occurring PI3Kα mutants. This compound is intended for research use only and not sold to patients.
- Potent mixed inhibitor of PI3Kα and PI3Kδ
- Selective against PI3Kβ and PI3Kγ
- Inhibits commonly occurring PI3Kα mutants
- Exhibits good solubility and permeability
- Low in vivo clearance and high bioavailability in rats and dogs