Learn More
Medchemexpress LLC AZD-8835 | 1620576-64-8 | 98.6% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY12869
Encompass_Preferred
AZD-8835 is a potent and selective inhibitor of PI3Kα and PI3Kδ, with IC50s of 6.2 and 5.7 nM respectively. This compound is intended for research use only.
  • Potent and selective inhibitor of PI3Kα and PI3Kδ
  • Purity of 98.6%
  • Available in solid form
  • Stable under recommended storage conditions
  • Suitable for research applications
Catalog No. 50-003-43072