Learn More
Medchemexpress LLC AZD 9272 | 327056-26-8 | 99.9% | 284.22 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY11025410MG
Encompass_Preferred
AZD 9272 is a selective, orally active, and brain penetrant mGluR5 antagonist. It demonstrates high selectivity over other mGlu receptors and is intended for research use only. This compound causes a concentration-dependent decrease in the intracellular Ca2+ response in mGluR5 expressing cell lines.
  • Selective mGluR5 antagonist
  • Orally active and brain penetrant
  • Shows >3900-fold selectivity over other mGlu receptors
  • Causes concentration-dependent decrease in intracellular Ca2+ response
  • Completely reverses glutamate-stimulated phosphatidyl inositol hydrolysis
Catalog No. 50-003-43115