Learn More
Medchemexpress LLC AZD-9574-acid | 2923687-90-3 | 99.4% | 415.39 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1609371MG
Encompass_Preferred
AZD-9574-acid (70D) is a PPAR-1 inhibitor that can be utilized for the synthesis of PROTAC.
  • PPAR-1 inhibitor
  • Can be used for the synthesis of PROTAC
Catalog No. 50-003-94354