Learn More
Medchemexpress LLC Azd-co-ph-peg4-ph-co-azd | 2569143-72-0 | MFCD00231029 | 96.0% | C26H28N2O8 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14180810MG
Encompass_Preferred
AZD-CO-Ph-PEG4-Ph-CO-AZD is a bis-β-lactam linker developed for synthesis of antibody-siRNA conjugates. It is supplied as a white to off-white solid (C26H28N2O8; MW 496.51; CAS 2569143-72-0) with reported purity of approximately 96%, and is soluble in DMSO with recommended formulations for in vivo use.
  • Bis-β-lactam linker suitable for antibody-siRNA conjugation.
  • High reported purity (~96%) supporting experimental reproducibility.
  • Solid form simplifies storage and handling.
  • Soluble in DMSO (50 mg/mL) for in vitro applications.
  • Includes recommended in vivo formulation options for dosing studies.
  • Available in small pack sizes appropriate for research use.
Catalog No. 50-002-60262