Learn More
TARGETMOL CHEMICALS INC AZD1208 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2300
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AZD1208 is a novel orally bioavailable highly selective PIM kinase inhibitor with single nanomolar potency against all three PIM kinases. purity: 98%
Catalog No. 50-257-7169
Edge