Learn More
Medchemexpress LLC HY-109566 10mg , AZD1390 CAS:2089288-03-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10956610MG
Encompass_Preferred
HY-109566 10mg AZD1390 CAS: 2089288-03-7 AZD1390 is a potent, highly selective, orally bioavailable, brain-penetrant ATM inhibitor with an IC50 of 0.78 nM in cell. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9930