Learn More
Selleck Chemical LLC AZD1480 10mM/1mL 935666-88-9
SDP
Supplier:  Selleck Chemical LLC S216210MM/1ML
Encompass_Preferred
AZD1480 is a novel ATP-competitive JAK2 inhibitor with IC50 of 0.26 nM in a cell-free assay, selectivity against JAK3 and Tyk2, and to a smaller extent against JAK1. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1001469