Learn More
TARGETMOL CHEMICALS INC AZD1940 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T302525MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AZD1940 (UNII-0J0035E9FT) is a high affinity CB(1)/CB(2) receptor agonist of the cannabinoid with oral activity. Application in the study of orofacial pain. purity: 99%
Catalog No. 50-258-0421