Learn More
Selleck Chemical LLC AZD3229 S8780-5MG
SDP
Supplier:  Selleck Chemical LLC S87805MG
Encompass_Preferred
AZD3229 is a potent pan-Kit (c-Kit) mutant inhibitor with potent single digit nM growth inhibition against a diverse panel of mutant Kit (c-Kit) driven Ba/F3 cell lines (GI50 1-50 nM) with good margin to KDR-driven effects It also inhibits PDGFR mutants (Tel-PDGFR Tel-PDGFR V561D/D842V)
Catalog No. 50-000-24485