Learn More
Selleck Chemical LLC Lanabecestat
Supplier: Selleck Chemical LLC S81932MG
Lanabecestat (AZD3293 LY3314814) is an oral beta-secretase 1 cleaving enzyme (BACE) inhibitor with an inhibitory constant Ki of 0 4 nM