Learn More
Medchemexpress LLC HY-112088 50mg Medchemexpress, AZD4573 CAS:2057509-72-3 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY11208850MG
Encompass_Preferred
Medchemexpress,HY-112088 50mg AZD4573 CAS:2057509-72-3 Formula:C22H28ClN5O2 AZD4573 is a CDK9 inhibitor with an IC50 of <3 nM extracted from patent US 20160376287 A1, example 14. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-7549