Learn More
Medchemexpress LLC HY-100653A 10mg , AZD5153 (6-Hydroxy-2-naphthoic acid) CAS:1869912-40-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY100653A10MG
Encompass_Preferred
HY-100653A 10mg AZD5153 (6-Hydroxy-2-naphthoic acid) CAS: 1869912-40-2 AZD5153 6-Hydroxy-2-naphthoic acid is the 6-Hydroxy-2-naphthoic acid of AZD5153. AZD5153 is a potent, selective, and orally available BET/BRD4 bromodomain inhibitor; disrupts BRD4 with an IC50 of 1.7 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9866