Learn More
Selleck Chemical LLC AZD5153 HNT salt
SDP
Supplier:  Selleck Chemical LLC S834425MG
Encompass_Preferred
AZD5153 6-hydroxy-2-naphthoic acid (HNT salt) is a potent selective and orally available BET/BRD4 bromodomain inhibitor with pKi of 8 3 for BRD4 AZD5153 inhibits the expression of Nuclear receptor binding SET domain protein 3 (NSD3) target genes NSD3 via H3K36me2 acts as an epigenetic deregulator to facilitate the expression of oncogenesis-promoting genes
Catalog No. 50-000-00653