Learn More
Medchemexpress LLC HY-132167 25mg , AZD5305 CAS:2589531-76-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13216725MG
Encompass_Preferred
HY-132167 25mg AZD5305 CAS: 2589531-76-8 AZD5305 is a potent, selective and oral active PARP inhibitor. AZD5305 is potent and efficacious in animal xenografts and PDX models. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0043