Learn More
Selleck Chemical LLC AZD5438 10mg 602306-29-6
SDP
Supplier:  Selleck Chemical LLC S262110MG
Encompass_Preferred
AZD5438 is a potent inhibitor of CDK1/2/9 with IC50 of 16 nM/6 nM/20 nM in cell-free assays. It is less potent to CDK5/6 and also inhibits GSK3?. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-877-8