Learn More
Medchemexpress LLC HY-12600 5mg Medchemexpress, AZD5582 CAS:1258392-53-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY126005MG
Encompass_Preferred
Medchemexpress, HY-12600 5mg AZD5582 CAS:1258392-53-8 AZD5582 is an antagonist of the inhibitor of apoptosis proteins (IAPs), which binds to the BIR3 domains cIAP1, cIAP2, and XIAP with IC50s of 15, 21, and 15 nM, respectively. AZD5582 induces apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8898