Learn More
Apexbio Technology LLC AZD6244 (Selumetinib) 606143-52-6 500mg
SDP
Supplier:  Apexbio Technology LLC A8207500
Encompass_Preferred
AZD6244 (Selumetinib CAS 606143-52-6) is a small-molecule inhibitor selectively targeting MEK1/2 components of the MAP kinase signaling cascade It inhibits MEK1 via a non-ATP-competitive mechanism with an IC50 of 14 1 nM leading to decreased phosphorylation of downstream targets ERK1/2 (IC50 10 3 nM cellular assays) AZD6244 exhibits minimal off-target activity against related kinases including MKK6 EGFR ErbB2 and B-Raf In tumor cell lines harboring activated Ras or B-Raf mutations AZD6244 reduces cell proliferation and inhibits growth in xenograft models Thus AZD6244 is useful for investigating MAP kinase pathway-driven malignancies
Catalog No. 50-000-62498