Learn More
Apexbio Technology LLC AZD6482, 5mg. Cas: 1173900-33-8 MFCD: MFCD16659062. PI3Kβ inhibitor,potent and selective
SDP
Supplier:  Apexbio Technology LLC A54785
Encompass_Preferred
AZD6428 is a potent, selective and ATP-competitive PI3K inhibitor. IC50 are 0.69, 13.6, 47.8 and 136 nM for PI 3-K, PI 3-K, PI 3-K and PI 3-K, respectively. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4954