Learn More
Apexbio Technology LLC AZD7545, 5mg. Cas: 252017-04-2 MFCD: MFCD25976792. PDHK inhibitor,potent and selective
SDP
Supplier:  Apexbio Technology LLC B12865
Encompass_Preferred
AZD7545 is a selective inhibitor of pyruvate dehydrogenase kinase 2 (PDHK2) with IC50 value of 6.4nM [1].PDH catalyzes the decarboxylation of pyruvate, converting it to acetyl-CoA. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5214