Learn More
Selleck Chemical LLC AZD8797-E1616-1G
SDP
Supplier:  Selleck Chemical LLC E16161G
Encompass_Preferred
AZD8797 (KAND567) is an allosteric non-competitive and orally active antagonist of the human receptor CX3CR1 It antagonizes CX3CR1 with a Ki value of 3 9 nM respectively It effects G-protein signaling and -arrestin recruitment
Catalog No. 50-313-6425