Learn More
Medchemexpress LLC HY-111269 5mg , AZD8848 CAS:866269-28-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1112695MG
Encompass_Preferred
HY-111269 5mg AZD8848 CAS: 866269-28-5 AZD8848 is a selective toll-like receptor 7 (TLR7) antedrug agonist which is developed for the research of asthma and allergic rhinitis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1288