Learn More
Selleck Chemical LLC AZD9496 5mg 1639042-08-2
SDP
Supplier:  Selleck Chemical LLC S83725MG
Encompass_Preferred
AZD9496 is an oral estrogen receptor inhibitor that blocks the growth of ER-positive and ESR1 mutant breast tumours in preclinical models. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6001