Learn More
Medchemexpress LLC AZD9496 deacrylic acid phenol | 2173404-70-9 | 96.6% | 388.43 | C22H23F3N2O | 10 MG

Supplier: Medchemexpress LLC HY4634010MG
AZD9496 deacrylic acid phenol is a small-molecule fragment derived from the AZD9496 ligand, used as the target protein ligand fragment in PROTAC ER degrader research. The compound is supplied as a solid for research use only and is characterized by its reported formula, molecular weight, and purity.
- Molecular formula: C22H23F3N2O.
- Molecular weight: 388.43.
- Reported purity: 96.6%.
- Appearance: light yellow to yellow solid.
- Available as small-scale research quantities.
- Storage: follow supplier recommendations for low-temperature storage under inert atmosphere when applicable.
- Intended use: research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.