Learn More
Vector Laboratories AZDye 568 NHS Ester (FP-1081) 100MG
SDP
Supplier:  Vector Laboratories FP1081100
Encompass
AZDye™ 568 NHS Ester (Alexa Fluor 568 NHS Ester equivalent) is an amine-reactive, orange fluorescent dye routinely used to label proteins or antibodies through the primary amines (Lys), amine-modified oligonucleotides, and other amine-containing biomolecules. AZDye™ 568 dye can be used for proteins labeling at high molar ratios without significantself-quenching, enabling brighter conjugates and more sensitive detection.
Catalog No. NC2948464