Learn More
Medchemexpress LLC Azelaic acid | 123-99-9 | MFCD00004432 | 98.0% | 100 G
SDP
Supplier:  Medchemexpress LLC HYB0704100G
Encompass_Preferred
Azelaic acid is a nine-carbon dicarboxylic acid with antimicrobial activity against *Propionibacterium acnes* and *Staphylococcus epidermidis* by inhibiting microbial cellular protein synthesis. It also has a hypopigmentation action due to its ability to scavenge free radicals.
  • Exhibits antimicrobial activity against *Propionibacterium acnes* and *Staphylococcus epidermidis*
  • Decreases intracellular reactive oxygen species (ROS) levels and increases antioxidant capacity
  • Suppresses B16, HMB2, and SK23 cell survival in a dose-dependent manner
  • Efficacious in treating moderate papulo-pustular rosacea
  • Suitable for laboratory centrifugation and storage
Catalog No. 50-004-08919